Assay Details
Target Gene Details
Entrez Gene ID: | 3512 |
Gene Name: | joining chain of multimeric IgA and IgM |
Gene Aliases: |
IGCJ, IGJ, JCH |
Location: |
Chr.4:70655541-70676901 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JCHAIN | XM_006714212.2 | XP_006714275.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829965 | Chr.4:70630001 - 70806591 on Build GRCh38 | Gain | JCHAIN UTP3 ENAM RUFY3 |
nsv1012495 | Chr.4:69979953 - 70947088 on Build GRCh38 | Gain | GRSF1 SMR3A FDCSP ODAM JCHAIN ENAM CSN1S2BP SMR3B CABS1 MUC7 HTN3 AMTN AMBN HTN1 PRR27 MOB1B STATH CSN1S2AP UTP3 RUFY3 CSN3 OPRPN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |