Assay Details
Target Gene Details
Entrez Gene ID: | 10941 |
Gene Name: | UDP glucuronosyltransferase family 2 member A1 complex locus |
Gene Aliases: |
UDPGT2A1 |
Location: |
Chr.4:69588417-69653249 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UGT2A1 | NM_001252274.2 | NP_001239203.2 | ||
NM_001252275.2 | NP_001239204.2 | |||
NM_001301239.1 | NP_001288168.1 | |||
NM_006798.4 | NP_006789.3 | |||
AJ006054.1 | CAB41974.1 | |||
AK304249.1 | ||||
AK314209.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3600919 | Chr.4:69621467 - 69651053 on Build GRCh38 | Loss | UGT2A2 UGT2A1 |
dgv5318n100 | Chr.4:69619810 - 69656037 on Build GRCh38 | Loss | UGT2A2 UGT2A1 |
nsv1009165 | Chr.4:69622101 - 69643600 on Build GRCh38 | Loss | UGT2A2 UGT2A1 |
dgv659n27 | Chr.4:69622642 - 69656037 on Build GRCh38 | Loss | UGT2A2 UGT2A1 |
esv3569511 | Chr.4:69621459 - 69650802 on Build GRCh38 | Loss | UGT2A2 UGT2A1 |
nsv594616 | Chr.4:69596324 - 69681535 on Build GRCh38 | Loss | UGT2A2 UGT2A1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |