Assay Details
Target Gene Details
Entrez Gene ID: | 60558 |
Gene Name: | GUF1 homolog, GTPase |
Gene Aliases: |
EF-4 |
Location: |
Chr.4:44678395-44700685 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GUF1 | NM_021927.2 | NP_068746.2 | ||
XM_005248122.3 | XP_005248179.1 | |||
XM_011513732.2 | XP_011512034.1 | |||
AK023282.1 | BAB14507.1 | |||
AK025248.1 | BAB15090.1 | |||
AK225314.1 | ||||
BC036768.1 | AAH36768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1013488 | Chr.4:44541791 - 44974106 on Build GRCh38 | Loss | GUF1 GNPDA2 YIPF7 |
nsv4319 | Chr.4:44671518 - 44716896 on Build GRCh38 | Deletion | GUF1 GNPDA2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |