Assay Details
Target Gene Details
Entrez Gene ID: | 102723704 |
Gene Name: | uncharacterized LOC102723704 |
Gene Aliases: |
- |
Location: |
Chr.4:102777037-102799575 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC102723704 | NR_125932.1 | |||
AK098632.1 |