Assay Details
Target Gene Details
Entrez Gene ID: | 132949 |
Gene Name: | aminoadipate-semialdehyde dehydrogenase |
Gene Aliases: |
ACSF4, LYS2, NRPS1098, NRPS998 |
Location: |
Chr.4:56337687-56387677 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1161086 | Chr.4:56310907 - 56510457 on Build GRCh38 | Duplication | LOC100506514 KIAA1211 AASDH ARL9 PAICS SRP72 PPAT |
nsv829940 | Chr.4:56206175 - 56396564 on Build GRCh38 | Gain | KIAA1211 AASDH LOC101928760 PPAT |
dgv5213n100 | Chr.4:56076751 - 56538345 on Build GRCh38 | Gain | LOC100506514 THEGL KIAA1211 AASDH ARL9 PAICS SRP72 LOC101928760 PPAT |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |