Assay Details
Target Gene Details
Entrez Gene ID: | 79804 |
Gene Name: | HAND2 antisense RNA 1 (head to head) |
Gene Aliases: |
DEIN, NBLA00301 |
Location: |
Chr.4:173528600-173591255 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAND2-AS1 | NR_136195.1 | |||
NR_136196.1 | ||||
AK054605.1 | ||||
AK123668.1 | ||||
DA863153.1 | ||||
DB292439.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2670690 | Chr.4:173545465 - 173555576 on Build GRCh38 | Deletion | HAND2-AS1 |
esv3603213 | Chr.4:173545438 - 173555636 on Build GRCh38 | Loss | HAND2-AS1 |