Assay Details
Target Gene Details
Entrez Gene ID: | 4887 |
Gene Name: | neuropeptide Y receptor Y2 |
Gene Aliases: |
NPY2-R |
Location: |
Chr.4:155173514-155217078 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPY2R | XM_005263033.4 | XP_005263090.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)