Assay Details
Target Gene Details
Entrez Gene ID: | 102723704 |
Gene Name: | uncharacterized LOC102723704 |
Gene Aliases: |
- |
Location: |
Chr.4:102777037-102799575 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC102723704 | NR_125932.1 | |||
AK098632.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526256 | Chr.4:102791432 - 102806853 on Build GRCh38 | Loss | LOC102723704 UBE2D3 |