Assay Details
Target Gene Details
Entrez Gene ID: | 8404 |
Gene Name: | SPARC like 1 |
Gene Aliases: |
MAST 9, MAST9, PIG33, SC1 |
Location: |
Chr.4:87473330-87529503 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPARCL1 | NM_001128310.2 | NP_001121782.1 | ||
NM_001291976.1 | NP_001278905.1 | |||
NM_001291977.1 | NP_001278906.1 | |||
NM_004684.5 | NP_004675.3 | |||
AK294335.1 | ||||
AK298039.1 | ||||
AK300956.1 | ||||
AK304494.1 | ||||
AK304628.1 | ||||
AK316533.1 | ||||
AL833430.1 | ||||
AY513278.1 | AAT08031.1 | |||
BC033721.1 | AAH33721.1 | |||
BX647713.1 | ||||
CA442598.1 | ||||
X82157.1 | CAA57650.1 | |||
X86693.1 | CAA60386.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3601276 | Chr.4:87422870 - 87515781 on Build GRCh38 | Loss | SPARCL1 NUDT9 |
esv3601278 | Chr.4:87455460 - 87496054 on Build GRCh38 | Loss | SPARCL1 NUDT9 |
nsv1011882 | Chr.4:87442807 - 87481325 on Build GRCh38 | Gain | SPARCL1 NUDT9 |
nsv1002789 | Chr.4:87318188 - 87685333 on Build GRCh38 | Loss | DSPP HSD17B11 SPARCL1 DMP1 NUDT9 HSD17B13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |