Assay Details
Target Gene Details
Entrez Gene ID: | 3815 |
Gene Name: | KIT proto-oncogene receptor tyrosine kinase |
Gene Aliases: |
C-Kit, CD117, PBT, SCFR |
Location: |
Chr.4:54657928-54740715 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KIT | NM_000222.2 | NP_000213.1 | ||
NM_001093772.1 | NP_001087241.1 | |||
XM_005265740.1 | XP_005265797.1 | |||
XM_005265741.1 | XP_005265798.1 | |||
XM_005265742.2 | XP_005265799.1 | |||
XM_017008178.1 | XP_016863667.1 | |||
XM_017008179.1 | XP_016863668.1 | |||
XM_017008180.1 | XP_016863669.1 | |||
AK304031.1 | ||||
BC071593.1 | AAH71593.1 | |||
DC376760.1 | ||||
EU826594.1 | ||||
GU983671.1 | ||||
HM015525.1 | ||||
HM015526.1 | ||||
KT326922.1 | ||||
X06182.1 | CAA29548.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526344 | Chr.4:54656321 - 55364952 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)