Assay Details
Target Gene Details
Entrez Gene ID: | 166647 |
Gene Name: | adhesion G protein-coupled receptor A3 |
Gene Aliases: |
GPR125, PGR21, TEM5L |
Location: |
Chr.4:22387374-22516058 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRA3 | NM_145290.3 | NP_660333.2 | ||
XM_005248137.4 | XP_005248194.1 | |||
XM_011513811.2 | XP_011512113.1 | |||
XM_017007834.1 | XP_016863323.1 | |||
AK027494.1 | ||||
AK095866.1 | ||||
AK299168.1 | ||||
AL832257.1 | ||||
AY181243.1 | AAO27355.1 | |||
BC035645.1 | AAH35645.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |