Assay Details
Target Gene Details
Entrez Gene ID: | 254827 |
Gene Name: | N-acetylated alpha-linked acidic dipeptidase-like 2 |
Gene Aliases: |
- |
Location: |
Chr.3:174439669-175809510 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Target Gene Details
Entrez Gene ID: | 100874244 |
Gene Name: | NAALADL2 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.3:175235092-175271096 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAALADL2-AS2 | NR_046713.1 | |||
BQ049755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829800 | Chr.3:175252630 - 175424883 on Build GRCh38 | Gain+Loss | NAALADL2 NAALADL2-AS2 MIR4789 |
nsv592550 | Chr.3:175143245 - 175354813 on Build GRCh38 | Loss | NAALADL2 NAALADL2-AS2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |