Assay Details
Target Gene Details
Entrez Gene ID: | 56852 |
Gene Name: | RAD18, E3 ubiquitin protein ligase |
Gene Aliases: |
RNF73 |
Location: |
Chr.3:8877196-8969249 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAD18 | NM_020165.3 | NP_064550.3 | ||
XM_017006873.1 | XP_016862362.1 | |||
AB035274.2 | BAA99284.1 | |||
AF169796.1 | AAF80856.1 | |||
AK023075.1 | BAB14392.1 | |||
AK222771.1 | BAD96491.1 | |||
AY004333.1 | AAF86618.1 | |||
BC001302.1 | AAH01302.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)