Assay Details
Target Gene Details
Entrez Gene ID: | 201595 |
Gene Name: | STT3B, catalytic subunit of the oligosaccharyltransferase complex |
Gene Aliases: |
CDG1X, SIMP, STT3-B |
Location: |
Chr.3:31532501-31637622 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STT3B | NM_178862.2 | NP_849193.1 | ||
XM_011533465.1 | XP_011531767.1 | |||
XM_017005857.1 | XP_016861346.1 | |||
XM_017005858.1 | XP_016861347.1 | |||
XM_017005859.1 | XP_016861348.1 | |||
AK074587.1 | ||||
AK311488.1 | ||||
AX799082.1 | CAE48447.1 | |||
AY074880.1 | AAL71884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain | CMTM8 TRIM71 CMTM6 LOC105377021 OSBPL10 GLB1 GPD1L STT3B LOC105377022 CMTM7 MIR548AY CCR4 TMPPE SUSD5 OSBPL10-AS1 DYNC1LI1 CNOT10 CRTAP ZNF860 |
nsv954458 | Chr.3:31535409 - 31660708 on Build GRCh38 | Duplication | STT3B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |