Assay Details
Target Gene Details
Entrez Gene ID: | 10438 |
Gene Name: | C1D nuclear receptor corepressor |
Gene Aliases: |
LRP1, Rrp47, SUN-CoR, SUNCOR, hC1D |
Location: |
Chr.2:68042200-68063027 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1D | NM_001190263.1 | NP_001177192.1 | ||
NM_001190265.1 | NP_001177194.1 | |||
NM_006333.3 | NP_006324.1 | |||
NM_173177.2 | NP_775269.1 | |||
AK290451.1 | ||||
BC005235.1 | AAH05235.1 | |||
BC009584.1 | AAH09584.1 | |||
BC016284.2 | AAH16284.1 | |||
BG505686.1 | ||||
DA900336.1 | ||||
X95592.1 | CAA64845.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834251 | Chr.2:68015596 - 68200603 on Build GRCh38 | Loss | C1D PPP3R1 WDR92 PNO1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |