Assay Details
Target Gene Details
Entrez Gene ID: | 129804 |
Gene Name: | fibulin 7 |
Gene Aliases: |
TM14 |
Location: |
Chr.2:112138385-112254888 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FBLN7 |
Target Gene Details
Entrez Gene ID: | 84524 |
Gene Name: | zinc finger CCCH-type containing 8 |
Gene Aliases: |
Fliz1, ZC3HDC8 |
Location: |
Chr.2:112215035-112255087 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZC3H8 | NM_032494.2 | NP_115883.2 | ||
XM_017005107.1 | XP_016860596.1 | |||
AF334161.1 | AAK13496.1 | |||
AK315329.1 | ||||
BC032001.1 | AAH32001.1 | |||
BG773518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss | PAFAH1B1P2 MIR4771-2 ACOXL MIR4435-2HG SNORD132 FBLN7 ANAPC1 TMEM87B MERTK ZC3H8 BCL2L11 MIR4435-2 BUB1 ZC3H6 LOC400997 |
nsv582703 | Chr.2:112162642 - 112342437 on Build GRCh38 | Loss | ZC3H8 ZC3H6 FBLN7 |
dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss | PAFAH1B1P2 MIR4771-2 ACOXL LOC105373553 MIR4435-2HG SNORD132 LOC100288570 FBLN7 LINC01106 ANAPC1 TMEM87B LOC105375809 MERTK LIMS4 ZC3H8 RGPD6 BCL2L11 MIR4435-2 BUB1 ZC3H6 LOC400997 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |