Assay Details
Target Gene Details
Entrez Gene ID: | 101928386 |
Gene Name: | uncharacterized LOC101928386 |
Gene Aliases: |
- |
Location: |
Chr.2:143937067-143964156 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928386 | NR_110237.1 |