Assay Details
Target Gene Details
Entrez Gene ID: | 10461 |
Gene Name: | MER proto-oncogene, tyrosine kinase |
Gene Aliases: |
MER, RP38, Tyro12, c-Eyk, c-mer |
Location: |
Chr.2:111898479-112039946 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MERTK | NM_006343.2 | NP_006334.2 | ||
XM_005263565.4 | XP_005263622.1 | |||
XM_005263568.4 | XP_005263625.1 | |||
XM_011510490.2 | XP_011508792.1 | |||
XM_017003164.1 | XP_016858653.1 | |||
XM_017003165.1 | XP_016858654.1 | |||
AK316584.1 | ||||
BC114917.1 | ||||
BC114918.1 | AAI14919.1 | |||
BX474860.1 | ||||
U08023.1 | AAB60430.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv998028 | Chr.2:111891895 - 111976438 on Build GRCh38 | Loss | MERTK |
esv3893093 | Chr.2:111912373 - 112100908 on Build GRCh38 | Gain | TMEM87B MERTK |
nsv582699 | Chr.2:110763166 - 112172487 on Build GRCh38 | Loss | LOC400997 TMEM87B FBLN7 PAFAH1B1P2 MIR4771-2 BCL2L11 MERTK ANAPC1 ACOXL MIR4435-2 MIR4435-2HG |
nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss | PAFAH1B1P2 BUB1 MIR4771-2 ZC3H6 ANAPC1 MIR4435-2HG LOC400997 ZC3H8 SNORD132 TMEM87B FBLN7 BCL2L11 MERTK ACOXL MIR4435-2 |
nsv526647 | Chr.2:111893739 - 111980985 on Build GRCh38 | Loss | MERTK |
esv2759083 | Chr.2:111142206 - 111989360 on Build GRCh38 | Gain+Loss | PAFAH1B1P2 MIR4771-2 BCL2L11 MERTK ANAPC1 MIR4435-2 MIR4435-2HG |
dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss | LOC100288570 LOC105373553 PAFAH1B1P2 BUB1 MIR4771-2 LINC01106 ZC3H6 ANAPC1 MIR4435-2HG RGPD6 LIMS4 LOC400997 ZC3H8 SNORD132 TMEM87B FBLN7 BCL2L11 LOC105375809 MERTK ACOXL MIR4435-2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |