Assay Details
Target Gene Details
Entrez Gene ID: | 6509 |
Gene Name: | solute carrier family 1 member 4 |
Gene Aliases: |
ASCT1, SATT, SPATCCM |
Location: |
Chr.2:64988445-65023865 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC1A4 | NM_001193493.1 | NP_001180422.1 | ||
NM_003038.4 | NP_003029.2 | |||
XM_006712079.2 | XP_006712142.1 | |||
AB026689.1 | BAA94861.1 | |||
AK223486.1 | BAD97206.1 | |||
AK295687.1 | ||||
AK298472.1 | ||||
AK313052.1 | ||||
AK315986.1 | ||||
BC026216.1 | AAH26216.1 | |||
BC072423.1 | AAH72423.1 | |||
CR990382.1 | ||||
DA218921.1 | ||||
L14595.1 | AAA02761.1 | |||
L19444.1 | AAA19438.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834246 | Chr.2:64981876 - 65176937 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1010909 | Chr.2:64985044 - 65027965 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)