Assay Details
Target Gene Details
Entrez Gene ID: | 5334 |
Gene Name: | phospholipase C like 1 |
Gene Aliases: |
PLCE, PLCL, PLDL1, PPP1R127, PRIP |
Location: |
Chr.2:197804702-198149884 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLCL1 | NM_006226.3 | NP_006217.3 | ||
XM_005246643.3 | XP_005246700.1 | |||
XM_011511351.2 | XP_011509653.1 | |||
AK127514.1 | ||||
AK302673.1 | ||||
DA193701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834506 | Chr.2:197723410 - 197881428 on Build GRCh38 | Gain |
![]() ![]() |
nsv460016 | Chr.2:197766990 - 197848507 on Build GRCh38 | Gain |
![]() ![]() |
nsv1009134 | Chr.2:197775024 - 197837700 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)