Assay Details
Target Gene Details
Entrez Gene ID: | 105374596 |
Gene Name: | uncharacterized LOC105374596 |
Gene Aliases: |
- |
Location: |
Chr.2:52370648-52390070 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105374596 | NR_135239.1 |
Target Gene Details
Entrez Gene ID: | 730100 |
Gene Name: | uncharacterized LOC730100 |
Gene Aliases: |
- |
Location: |
Chr.2:51032601-52407917 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC730100 | NR_135237.1 | |||
AK127244.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv581928 | Chr.2:52345029 - 52429456 on Build GRCh38 | Loss | LOC730100 LOC105374596 |
nsv834025 | Chr.2:52287821 - 52465911 on Build GRCh38 | Gain | LOC730100 LOC105374596 |
esv3583824 | Chr.2:52372222 - 52511051 on Build GRCh38 | Loss | LOC730100 LOC105374596 |