Assay Details
Target Gene Details
Entrez Gene ID: | 10018 |
Gene Name: | BCL2 like 11 |
Gene Aliases: |
BAM, BIM, BOD |
Location: |
Chr.2:111120914-111168445 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv582699 | Chr.2:110763166 - 112172487 on Build GRCh38 | Loss | ANAPC1 FBLN7 ACOXL MIR4435-2 MIR4435-2HG TMEM87B LOC400997 BCL2L11 PAFAH1B1P2 MERTK MIR4771-2 |
nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss | ANAPC1 ACOXL SNORD132 LOC400997 PAFAH1B1P2 ZC3H6 BUB1 FBLN7 MIR4435-2 MIR4435-2HG TMEM87B BCL2L11 ZC3H8 MERTK MIR4771-2 |
nsv834331 | Chr.2:111054562 - 111247101 on Build GRCh38 | Loss | ACOXL MIR4435-2HG LOC400997 BCL2L11 |
nsv428402 | Chr.2:111142206 - 111905020 on Build GRCh38 | Loss | ANAPC1 MIR4435-2 MIR4435-2HG BCL2L11 PAFAH1B1P2 MERTK MIR4771-2 |
dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss | ANAPC1 ACOXL SNORD132 LOC105375809 LOC105373553 LOC400997 PAFAH1B1P2 LINC01106 ZC3H6 BUB1 FBLN7 MIR4435-2 RGPD6 MIR4435-2HG TMEM87B BCL2L11 LIMS4 ZC3H8 LOC100288570 MERTK MIR4771-2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |