Assay Details
Target Gene Details
Entrez Gene ID: | 4664 |
Gene Name: | NGFI-A binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.2:190648860-190692766 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAB1 | NM_001321312.1 | NP_001308241.1 | ||
NM_001321313.1 | NP_001308242.1 | |||
NM_005966.3 | NP_005957.2 | |||
XM_005246582.1 | XP_005246639.1 | |||
XM_005246583.1 | XP_005246640.1 | |||
XM_011511219.2 | XP_011509521.1 | |||
XM_017004170.1 | XP_016859659.1 | |||
XM_017004171.1 | XP_016859660.1 | |||
XM_017004172.1 | XP_016859661.1 | |||
XM_017004173.1 | XP_016859662.1 | |||
XM_017004174.1 | XP_016859663.1 | |||
XM_017004175.1 | XP_016859664.1 | |||
XM_017004176.1 | XP_016859665.1 | |||
AK292446.1 | ||||
BC035724.1 | AAH35724.1 | |||
DB457461.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3082 | Chr.2:190627513 - 190688068 on Build GRCh38 | Insertion | NAB1 |
nsv834492 | Chr.2:190574138 - 190781895 on Build GRCh38 | Gain | NEMP2 NAB1 |
dgv1200e212 | Chr.2:190638342 - 190674560 on Build GRCh38 | Loss | NAB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |