Assay Details
Target Gene Details
Entrez Gene ID: | 129804 |
Gene Name: | fibulin 7 |
Gene Aliases: |
TM14 |
Location: |
Chr.2:112138385-112254888 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FBLN7 | NM_001128165.1 | NP_001121637.1 | ||
NM_153214.2 | NP_694946.2 | |||
XM_006712260.3 | XP_006712323.1 | |||
XM_011510585.2 | XP_011508887.1 | |||
XM_011510587.2 | XP_011508889.1 | |||
XM_017003317.1 | XP_016858806.1 | |||
XM_017003318.1 | XP_016858807.1 | |||
XM_017003319.1 | XP_016858808.1 | |||
AK094759.1 | BAC04416.1 | |||
BC035784.1 | ||||
BC126986.1 | ||||
BC126987.1 | ||||
CR933697.1 | CAI46168.1 | |||
DA201691.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv582699 | Chr.2:110763166 - 112172487 on Build GRCh38 | Loss | MIR4435-2 MIR4771-2 FBLN7 ANAPC1 TMEM87B MIR4435-2HG MERTK BCL2L11 PAFAH1B1P2 ACOXL LOC400997 |
nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss | SNORD132 MIR4435-2 MIR4771-2 FBLN7 MIR4435-2HG ZC3H8 MERTK PAFAH1B1P2 ZC3H6 ANAPC1 TMEM87B BCL2L11 ACOXL LOC400997 BUB1 |
dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss | SNORD132 MIR4435-2 MIR4771-2 FBLN7 LOC100288570 MIR4435-2HG ZC3H8 MERTK PAFAH1B1P2 LINC01106 ZC3H6 LIMS4 RGPD6 ANAPC1 LOC105375809 TMEM87B LOC105373553 BCL2L11 ACOXL LOC400997 BUB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |