Assay Details
Target Gene Details
Entrez Gene ID: | 343066 |
Gene Name: | arylacetamide deacetylase like 4 |
Gene Aliases: |
- |
Location: |
Chr.1:12644547-12667086 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AADACL4 | NM_001013630.1 | NP_001013652.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3585236 | Chr.1:12642968 - 12652029 on Build GRCh38 | Loss | AADACL4 |
nsv1011540 | Chr.1:12601623 - 12832443 on Build GRCh38 | Loss | PRAMEF11 PRAMEF1 C1orf158 AADACL4 DHRS3 PRAMEF12 AADACL3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |