Assay Details
Target Gene Details
Entrez Gene ID: | 3716 |
Gene Name: | Janus kinase 1 |
Gene Aliases: |
JAK1A, JAK1B, JTK3 |
Location: |
Chr.1:64833223-65067746 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JAK1 | NM_001321852.1 | NP_001308781.1 | ||
NM_001321853.1 | NP_001308782.1 | |||
NM_001321854.1 | NP_001308783.1 |
Target Gene Details
Entrez Gene ID: | 101927084 |
Gene Name: | long intergenic non-protein coding RNA 1359 |
Gene Aliases: |
- |
Location: |
Chr.1:64979577-65002476 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01359 | NR_119383.1 | |||
BI825893.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1165 | Chr.1:64997218 - 65041683 on Build GRCh38 | Deletion | LINC01359 JAK1 |
esv2676423 | Chr.1:64977641 - 65405798 on Build GRCh38 | Deletion | LOC107983962 DNAJC6 MIR101-1 LINC01359 MIR3671 AK4 JAK1 |
nsv830059 | Chr.1:64918047 - 65074478 on Build GRCh38 | Loss | LOC107983962 MIR101-1 LINC01359 MIR3671 JAK1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |