Assay Details
Target Gene Details
Entrez Gene ID: | 7107 |
Gene Name: | G protein-coupled receptor 137B |
Gene Aliases: |
TM7SF1 |
Location: |
Chr.1:236142502-236211059 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR137B | NM_003272.3 | NP_003263.1 | ||
XM_017002209.1 | XP_016857698.1 | |||
XM_017002210.1 | XP_016857699.1 | |||
AF027826.1 | AAC39669.1 | |||
AK223632.1 | BAD97352.1 | |||
AK295408.1 | ||||
AK301552.1 | ||||
AK312908.1 | ||||
AY242135.1 | AAO92302.1 | |||
BC012100.1 | AAH12100.1 | |||
CR541770.1 | CAG46569.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv590n100 | Chr.1:236139422 - 236267713 on Build GRCh38 | Gain | GPR137B ERO1B |
nsv1005877 | Chr.1:235912335 - 237038508 on Build GRCh38 | Gain | EDARADD MT1HL1 HEATR1 LGALS8-AS1 GPR137B NID1 LGALS8 ACTN2 ERO1B MTR |
nsv468428 | Chr.1:236121000 - 236194091 on Build GRCh38 | Loss | GPR137B |
nsv832969 | Chr.1:236111012 - 236311443 on Build GRCh38 | Gain | GPR137B ERO1B |
nsv549394 | Chr.1:235876577 - 236629706 on Build GRCh38 | Gain | EDARADD HEATR1 LGALS8-AS1 GPR137B NID1 LGALS8 LYST ERO1B |
nsv1012436 | Chr.1:235860657 - 236648100 on Build GRCh38 | Loss | EDARADD HEATR1 LGALS8-AS1 GPR137B NID1 LGALS8 LYST ERO1B |