Assay Details
Target Gene Details
Entrez Gene ID: | 5563 |
Gene Name: | protein kinase AMP-activated catalytic subunit alpha 2 |
Gene Aliases: |
AMPK, AMPK2, AMPKa2, PRKAA |
Location: |
Chr.1:56645317-56715335 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRKAA2 | NM_006252.3 | NP_006243.2 | ||
XM_017001692.1 | XP_016857181.1 | |||
XM_017001693.1 | XP_016857182.1 | |||
XM_017001694.1 | XP_016857183.1 | |||
AK314011.1 | ||||
BC069680.1 | AAH69680.1 | |||
BC069740.1 | AAH69740.1 | |||
BC069823.1 | AAH69823.1 | |||
DA710012.1 | ||||
EF056019.1 | ||||
FJ200485.1 | ||||
FJ200486.1 | ||||
HQ628625.1 | ||||
U06454.1 | AAA64745.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv523032 | Chr.1:56651236 - 56783885 on Build GRCh38 | Gain | C1orf168 PRKAA2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |