Assay Details
Target Gene Details
Entrez Gene ID: | 64218 |
Gene Name: | semaphorin 4A |
Gene Aliases: |
CORD10, RP35, SEMAB, SEMB |
Location: |
Chr.1:156149673-156177751 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SEMA4A | NM_001193301.1 | NP_001180230.1 | ||
XM_011509871.2 | XP_011508173.1 | |||
XM_011509872.2 | XP_011508174.1 | |||
XM_011509873.2 | XP_011508175.1 | |||
XM_011509876.2 | XP_011508178.1 | |||
XM_011509878.2 | XP_011508180.1 | |||
XM_011509879.2 | XP_011508181.1 | |||
XM_017002057.1 | XP_016857546.1 | |||
AY358531.1 | AAQ88895.1 | |||
DA409501.1 | ||||
DA508826.1 | ||||
DC332598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509513 | Chr.1:155932322 - 156178790 on Build GRCh38 | Insertion | LMNA ARHGEF2 LAMTOR2 RAB25 SSR2 SEMA4A UBQLN4 MEX3A KIAA0907 MIR6738 RXFP4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |