Assay Details
Target Gene Details
Entrez Gene ID: | 5293 |
Gene Name: | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta |
Gene Aliases: |
APDS, IMD14, P110DELTA, PI3K, p110D |
Location: |
Chr.1:9629889-9729114 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIK3CD | NM_005026.3 | NP_005017.3 | ||
XM_006710689.2 | XP_006710752.1 | |||
XM_017001477.1 | XP_016856966.1 | |||
XM_017001478.1 | XP_016856967.1 | |||
XM_017001479.1 | XP_016856968.1 | |||
XM_017001480.1 | XP_016856969.1 | |||
XM_017001481.1 | XP_016856970.1 | |||
AK314191.1 | ||||
BC150297.1 | ||||
CN313981.1 | ||||
U86453.1 | AAC25677.1 | |||
Y10055.2 | CAA71149.2 |
Target Gene Details
Entrez Gene ID: | 101929074 |
Gene Name: | PIK3CD antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.1:9672426-9687574 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIK3CD-AS2 | NR_126366.1 | |||
NR_126367.1 | ||||
AA341665.1 | ||||
AL523099.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3585185 | Chr.1:9670031 - 9692162 on Build GRCh38 | Gain | PIK3CD PIK3CD-AS2 |
nsv508847 | Chr.1:9612137 - 9678831 on Build GRCh38 | Insertion | PIK3CD PIK3CD-AS2 TMEM201 LOC107985734 PIK3CD-AS1 |
nsv832203 | Chr.1:9666574 - 9800151 on Build GRCh38 | Loss | PIK3CD PIK3CD-AS2 CLSTN1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |