Assay Details
Target Gene Details
Entrez Gene ID: | 5052 |
Gene Name: | peroxiredoxin 1 |
Gene Aliases: |
MSP23, NKEF-A, NKEFA, PAG, PAGA, PAGB, PRX1, PRXI, TDPX2 |
Location: |
Chr.1:45511035-45522890 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRDX1 | NM_001202431.1 | NP_001189360.1 | ||
NM_002574.3 | NP_002565.1 | |||
NM_181696.2 | NP_859047.1 | |||
NM_181697.2 | NP_859048.1 | |||
AK131049.1 | ||||
AK311898.1 | ||||
AV691526.1 | ||||
BC007063.1 | AAH07063.1 | |||
BC021683.1 | AAH21683.1 | |||
BM740381.1 | ||||
BM762399.1 | ||||
BM787105.1 | ||||
BM827394.1 | ||||
BU564066.1 | ||||
CD638470.1 | ||||
L19184.1 | AAA50464.1 | |||
X67951.1 | CAA48137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain | NASP CCDC17 RPS15AP10 MAST2 AKR1A1 TESK2 IPP CCDC163P GPBP1L1 MMACHC PIK3R3 PRDX1 TMEM69 |
nsv829726 | Chr.1:45431909 - 45637514 on Build GRCh38 | Loss | NASP CCDC17 CCDC163P GPBP1L1 MMACHC PRDX1 AKR1A1 TESK2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |