Assay Details
Target Gene Details
Entrez Gene ID: | 7049 |
Gene Name: | transforming growth factor beta receptor 3 |
Gene Aliases: |
BGCAN, betaglycan |
Location: |
Chr.1:91680343-91906002 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGFBR3 | NM_001195683.1 | NP_001182612.1 | ||
NM_001195684.1 | NP_001182613.1 | |||
NM_003243.4 | NP_003234.2 | |||
NR_036634.1 | ||||
XM_006710867.2 | XP_006710930.1 | |||
XM_006710868.2 | XP_006710931.1 | |||
AJ251961.1 | CAB64374.1 | |||
AK225799.1 | ||||
AK291345.1 | ||||
BC126116.1 | ||||
BC136295.1 | ||||
HG764816.1 | ||||
L07594.1 | AAA67061.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2763581 | Chr.1:91603410 - 91738246 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)