Assay Details
Target Gene Details
Entrez Gene ID: | 9202 |
Gene Name: | zinc finger MYM-type containing 4 |
Gene Aliases: |
CDIR, MYM, ZNF198L3, ZNF262 |
Location: |
Chr.1:35268694-35421944 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZMYM4 | NM_005095.2 | NP_005086.2 | ||
XM_005271328.4 | XP_005271385.1 | |||
XM_005271331.2 | XP_005271388.1 | |||
XM_011542423.2 | XP_011540725.1 | |||
XM_011542425.2 | XP_011540727.1 | |||
XM_017002803.1 | XP_016858292.1 | |||
XM_017002804.1 | XP_016858293.1 | |||
AK308320.1 | ||||
BC127113.1 | ||||
BC127114.1 | ||||
BG499626.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |