Assay Details
Target Gene Details
Entrez Gene ID: | 83417 |
Gene Name: | Fc receptor like 4 |
Gene Aliases: |
CD307d, FCRH4, IGFP2, IRTA1 |
Location: |
Chr.1:157573749-157598080 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCRL4 | NM_031282.2 | NP_112572.1 | ||
XM_011510034.1 | XP_011508336.1 | |||
AF329490.1 | AAL23900.1 | |||
AF329492.1 | AAL23902.1 | |||
AF343659.1 | AAK31322.1 | |||
AF343660.1 | AAK31323.1 | |||
AF343661.1 | AAK31324.1 | |||
AF397452.1 | AAK93970.1 | |||
BC125173.1 | ||||
BC125174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2718507 | Chr.1:157533583 - 157635387 on Build GRCh38 | Deletion | FCRL5 FCRL4 |
nsv3154 | Chr.1:157562046 - 157596752 on Build GRCh38 | Insertion | FCRL4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |