Assay Details
Target Gene Details
Entrez Gene ID: | 10158 |
Gene Name: | PDZK1 interacting protein 1 |
Gene Aliases: |
DD96, MAP17, SPAP |
Location: |
Chr.1:47183589-47190099 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDZK1IP1 | NM_005764.3 | NP_005755.1 | ||
BC012303.1 | AAH12303.1 | |||
BM924665.1 | ||||
CR450304.1 | CAG29300.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950709 | Chr.1:47173129 - 47191428 on Build GRCh38 | Deletion | PDZK1IP1 LINC00853 |