Assay Details
Target Gene Details
Entrez Gene ID: | 23013 |
Gene Name: | spen family transcriptional repressor |
Gene Aliases: |
HIAA0929, MINT, RBM15C, SHARP |
Location: |
Chr.1:15847864-15940455 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPEN | NM_015001.2 | NP_055816.2 | ||
AF356524.1 | AAK52750.1 | |||
AK127577.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833647 | Chr.1:15794042 - 15952334 on Build GRCh38 | Loss | FLJ37453 ZBTB17 UQCRHL SPEN |
dgv37n100 | Chr.1:15823408 - 15924038 on Build GRCh38 | Gain | FLJ37453 SPEN |
dgv38n100 | Chr.1:15839166 - 15924038 on Build GRCh38 | Gain | FLJ37453 SPEN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |