Assay Details
Target Gene Details
Entrez Gene ID: | 164118 |
Gene Name: | tetratricopeptide repeat domain 24 |
Gene Aliases: |
- |
Location: |
Chr.1:156579727-156587717 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TTC24 | NM_001105669.2 | NP_001099139.2 |