Assay Details
Target Gene Details
Entrez Gene ID: | 100873946 |
Gene Name: | VAV3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:107964443-107994607 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VAV3-AS1 | NR_046653.1 | |||
CR744964.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1011093 | Chr.1:107774643 - 108219390 on Build GRCh38 | Gain | SLC25A24 VAV3 VAV3-AS1 MIR7852 |
dgv248n100 | Chr.1:107385172 - 108286989 on Build GRCh38 | Gain | SLC25A24 VAV3 VAV3-AS1 NTNG1 NBPF4 MIR7852 |
nsv524680 | Chr.1:107776457 - 108212812 on Build GRCh38 | Gain | SLC25A24 VAV3 VAV3-AS1 MIR7852 |