Assay Details
Target Gene Details
Entrez Gene ID: | 29937 |
Gene Name: | neudesin neurotrophic factor |
Gene Aliases: |
CIR2, SCIRP10, SPUF |
Location: |
Chr.1:212432887-212446379 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NENF | NM_013349.4 | NP_037481.1 | ||
NR_026598.1 | ||||
AB126219.1 | BAD72063.1 | |||
AF049672.1 | ||||
AF173937.1 | AAD51419.1 | |||
AK074431.1 | ||||
AK223135.1 | BAD96855.1 | |||
AY762102.1 | ||||
BC008823.2 | AAH08823.1 | |||
DB502983.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv549139 | Chr.1:212158952 - 212703560 on Build GRCh38 | Gain | PPP2R5A LOC101929565 LOC101929541 FAM71A BATF3 TMEM206 SNORA16B ATF3 NENF |
nsv527863 | Chr.1:212444081 - 212527276 on Build GRCh38 | Loss | NENF |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |