Assay Details
Target Gene Details
Entrez Gene ID: | 4904 |
Gene Name: | Y-box binding protein 1 |
Gene Aliases: |
BP-8, CBF-A, CSDA2, CSDB, DBPB, EFI-A, MDR-NF1, NSEP-1, NSEP1, YB-1, YB1 |
Location: |
Chr.1:42682235-42703803 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YBX1 | NM_004559.4 | NP_004550.2 | ||
NR_132737.1 | ||||
AK309996.1 | ||||
AK311708.1 | ||||
BC000064.2 | AAH00064.1 | |||
BC002411.2 | AAH02411.1 | |||
BC015208.1 | AAH15208.1 | |||
BC018393.1 | ||||
BC038384.1 | AAH38384.1 | |||
BC065571.1 | AAH65571.1 | |||
BC070084.1 | AAH70084.1 | |||
BC071708.1 | AAH71708.1 | |||
BC072014.1 | AAH72014.1 | |||
BC090038.1 | AAH90038.1 | |||
BC098435.1 | AAH98435.1 | |||
BC106045.1 | AAI06046.1 | |||
CN353145.1 | ||||
J03827.1 | AAA61308.1 | |||
L28809.1 | AAA20871.1 | |||
M24070.1 | AAA35750.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3577249 | Chr.1:42576524 - 42885152 on Build GRCh38 | Gain | LOC105378683 SVBP YBX1 CLDN19 ERMAP CCDC30 P3H1 PPIH LOC100129924 LOC339539 C1orf50 ZNF691 |
nsv829682 | Chr.1:42589429 - 42773568 on Build GRCh38 | Loss | YBX1 CLDN19 CCDC30 P3H1 PPIH C1orf50 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |