Assay Details
Target Gene Details
Entrez Gene ID: | 63036 |
Gene Name: | chymotrypsin like elastase family member 2A |
Gene Aliases: |
ELA2A, PE-1 |
Location: |
Chr.1:15456728-15472091 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CELA2A | NM_033440.2 | NP_254275.1 | ||
AK312198.1 | ||||
BC007031.1 | AAH07031.1 | |||
BC069331.1 | AAH69331.1 | |||
BC069432.1 | AAH69432.1 | |||
BP324428.1 | ||||
CR450291.1 | CAG29287.1 | |||
D00236.1 | BAA00165.1 | |||
M16631.1 | AAA52374.1 | |||
M16652.1 | AAA52380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2744308 | Chr.1:15455456 - 15475909 on Build GRCh38 | Deletion | CELA2A |
esv3548899 | Chr.1:15454506 - 15475973 on Build GRCh38 | Deletion | CELA2A |
nsv833536 | Chr.1:15323505 - 15528830 on Build GRCh38 | Loss | FHAD1 DNAJC16 LOC101927441 CELA2B EFHD2 CELA2A CASP9 CTRC |
esv2761582 | Chr.1:15457276 - 15475000 on Build GRCh38 | Loss | CELA2A |
nsv1114929 | Chr.1:15455856 - 15475193 on Build GRCh38 | Deletion | CELA2A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |