Assay Details
Target Gene Details
Entrez Gene ID: | 8880 |
Gene Name: | far upstream element binding protein 1 |
Gene Aliases: |
FBP, FUBP, hDH V |
Location: |
Chr.1:77944055-77979494 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FUBP1 | NM_001303433.1 | NP_001290362.1 | ||
NM_003902.4 | NP_003893.2 | |||
NR_130152.1 | ||||
XM_011542391.2 | XP_011540693.1 | |||
XM_011542392.2 | XP_011540694.1 | |||
XM_011542393.2 | XP_011540695.1 | |||
XM_017002739.1 | XP_016858228.1 | |||
XM_017002740.1 | XP_016858229.1 | |||
XM_017002741.1 | XP_016858230.1 | |||
XM_017002742.1 | XP_016858231.1 | |||
XM_017002743.1 | XP_016858232.1 | |||
XM_017002744.1 | XP_016858233.1 | |||
AB209366.1 | BAD92603.1 | |||
AK300032.1 | ||||
AK301581.1 | ||||
AK303568.1 | ||||
BC017247.2 | AAH17247.1 | |||
U05040.2 | AAA17976.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv518855 | Chr.1:77950984 - 77990671 on Build GRCh38 | Loss | FUBP1 DNAJB4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |