Assay Details
Target Gene Details
Entrez Gene ID: | 83593 |
Gene Name: | Ras association domain family member 5 |
Gene Aliases: |
Maxp1, NORE1, NORE1A, NORE1B, RAPL, RASSF3 |
Location: |
Chr.1:206507530-206589284 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RASSF5 | NM_182663.3 | NP_872604.1 | ||
NM_182664.3 | NP_872605.1 | |||
AB209145.1 | BAD92382.1 | |||
AK289861.1 | ||||
AK291268.1 | ||||
AY062002.1 | AAL40388.1 | |||
AY062003.1 | AAL40389.1 | |||
BC004270.1 | AAH04270.1 | |||
BC007203.1 | AAH07203.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)