Assay Details
Target Gene Details
Entrez Gene ID: | 9249 |
Gene Name: | dehydrogenase/reductase 3 |
Gene Aliases: |
DD83.1, RDH17, Rsdr1, SDR1, SDR16C1, retSDR1 |
Location: |
Chr.1:12567910-12618361 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DHRS3 | NM_001319225.1 | NP_001306154.1 | ||
NM_001324370.1 | NP_001311299.1 | |||
NM_004753.6 | NP_004744.2 | |||
XM_006711036.1 | XP_006711099.1 | |||
XM_017002809.1 | XP_016858298.1 | |||
AF061741.1 | AAC63263.1 | |||
AK222593.1 | BAD96313.1 | |||
AK225057.1 | ||||
AK309282.1 | ||||
AK310202.1 | ||||
AK312961.1 | ||||
AY358093.1 | AAQ88460.1 | |||
BC002730.2 | AAH02730.1 | |||
BX648476.1 | CAI46033.1 | |||
DQ426869.1 | ||||
DQ426870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521737 | Chr.1:12539396 - 12624831 on Build GRCh38 | Loss |
![]() ![]() |
nsv545443 | Chr.1:12565072 - 12624831 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)