Assay Details
Target Gene Details
Entrez Gene ID: | 5511 |
Gene Name: | protein phosphatase 1 regulatory subunit 8 |
Gene Aliases: |
ARD-1, ARD1, NIPP-1, NIPP1, PRO2047 |
Location: |
Chr.1:27830741-27851672 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R8 | NM_002713.3 | NP_002704.1 | ||
NM_014110.4 | NP_054829.2 | |||
NM_138558.2 | NP_612568.1 | |||
XM_017001594.1 | XP_016857083.1 | |||
AF061958.1 | AAD31541.1 | |||
AF061959.1 | AAD31542.1 | |||
AF126488.1 | AAD22486.1 | |||
AK292077.1 | ||||
AK297771.1 | ||||
AK304619.1 | ||||
AK307570.1 | ||||
AK313814.1 | ||||
AX775985.1 | CAE11692.1 | |||
BC001597.1 | AAH01597.1 | |||
BC013360.2 | AAH13360.1 | |||
BC093017.1 | AAH93017.1 | |||
BI459680.1 | ||||
CR450309.1 | CAG29305.1 | |||
DQ223931.1 | ABB04281.1 | |||
U14575.1 | AAA64749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509069 | Chr.1:27817460 - 27905956 on Build GRCh38 | Insertion | SCARNA1 PPP1R8 STX12 THEMIS2 RPA2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |