Assay Details
Target Gene Details
Entrez Gene ID: | 6464 |
Gene Name: | SHC adaptor protein 1 |
Gene Aliases: |
SHC, SHCA |
Location: |
Chr.1:154962298-154974492 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SHC1 | NM_001130041.1 | NP_001123513.1 | ||
NM_001202859.1 | NP_001189788.1 | |||
NM_003029.4 | NP_003020.2 | |||
XM_005245451.4 | XP_005245508.1 | |||
XM_011509897.1 | XP_011508199.1 | |||
XM_011509898.2 | XP_011508200.1 | |||
XM_017002083.1 | XP_016857572.1 | |||
AK130014.1 | ||||
AK292143.1 | ||||
AK315842.1 | ||||
BC014158.2 | AAH14158.1 | |||
X68148.1 | CAA48251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)