Assay Details
Target Gene Details
Entrez Gene ID: | 284677 |
Gene Name: | chromosome 1 open reading frame 204 |
Gene Aliases: |
- |
Location: |
Chr.1:159834474-159855347 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1orf204 | NM_001134233.1 | NP_001127705.1 | ||
AK096506.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005195 | Chr.1:159482969 - 159940547 on Build GRCh38 | Gain | IGSF9 C1orf204 DUSP23 VSIG8 OR10J5 MIR4259 CRP FCRL6 CFAP45 TAGLN2 SLAMF8 APCS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |