Assay Details
Target Gene Details
Entrez Gene ID: | 11332 |
Gene Name: | acyl-CoA thioesterase 7 |
Gene Aliases: |
ACH1, ACT, BACH, CTE-II, LACH, LACH1, hBACH |
Location: |
Chr.1:6264272-6393766 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACOT7 | NM_007274.3 | NP_009205.3 | ||
NM_181864.2 | NP_863654.1 | |||
NM_181865.2 | NP_863655.1 | |||
NM_181866.2 | NP_863656.1 | |||
AB074415.1 | BAC20174.1 | |||
AB074416.1 | BAC20175.1 | |||
AB074417.1 | BAC20176.1 | |||
AB074418.1 | BAC20177.1 | |||
AB074419.1 | BAC20178.1 | |||
AK057168.1 | ||||
AK289572.1 | ||||
AK290097.1 | ||||
AK291583.1 | ||||
AK292202.1 | ||||
AK300831.1 | ||||
BC017365.2 | ||||
BT006888.1 | AAP35534.1 | |||
D88894.2 | BAA24350.1 | |||
U91316.1 | AAB61211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv965 | Chr.1:6253120 - 6297822 on Build GRCh38 | Deletion | GPR153 ACOT7 |
nsv526371 | Chr.1:6247195 - 6307159 on Build GRCh38 | Loss | GPR153 ACOT7 |
nsv545239 | Chr.1:6262876 - 6289815 on Build GRCh38 | Loss | ACOT7 |
nsv508825 | Chr.1:6221275 - 6286549 on Build GRCh38 | Insertion | GPR153 ICMT LINC00337 RNF207 HES3 ACOT7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |