Assay Details
Target Gene Details
Entrez Gene ID: | 28982 |
Gene Name: | feline leukemia virus subgroup C cellular receptor 1 |
Gene Aliases: |
AXPC1, FLVCR, MFSD7B, PCA, PCARP |
Location: |
Chr.1:212858255-212899363 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FLVCR1 | NM_014053.3 | NP_054772.1 | ||
XM_011509446.2 | XP_011507748.1 | |||
XM_011509447.2 | XP_011507749.1 | |||
XM_011509448.1 | XP_011507750.1 | |||
AF118637.1 | AAD45243.1 | |||
AK001419.1 | BAA91679.1 | |||
AK314481.1 | ||||
BC048312.1 | AAH48312.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv572n100 | Chr.1:212856472 - 212899671 on Build GRCh38 | Gain |
![]() ![]() |
dgv571n100 | Chr.1:212831823 - 212896221 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1008383 | Chr.1:212852204 - 212941552 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)