Assay Details
Target Gene Details
Entrez Gene ID: | 115350 |
Gene Name: | Fc receptor like 1 |
Gene Aliases: |
CD307a, FCRH1, IFGP1, IRTA5 |
Location: |
Chr.1:157792249-157820150 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCRL1 | XM_005244866.2 | 11 | 4234 | XP_005244923.1 |
XM_005244867.2 | 10 | 4127 | XP_005244924.1 | |
XM_005244869.3 | 9 | 3969 | XP_005244926.1 | |
XM_011509125.1 | 12 | 4242 | XP_011507427.1 | |
XM_011509126.1 | 12 | 4239 | XP_011507428.1 | |
XM_011509127.2 | 10 | 4236 | XP_011507429.1 | |
XM_011509128.1 | 11 | 4246 | XP_011507430.1 | |
XM_011509129.1 | 11 | 4243 | XP_011507431.1 | |
XM_011509130.1 | 11 | 4231 | XP_011507432.1 | |
XM_011509133.2 | 11 | 4098 | XP_011507435.1 | |
XM_011509134.2 | 10 | 4073 | XP_011507436.1 | |
XM_011509135.2 | 10 | 3990 | XP_011507437.1 | |
XM_011509137.2 | 8 | 3618 | XP_011507439.1 | |
XM_017000227.1 | 10 | 6240 | XP_016855716.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |